Anti-CCR8 Rabbit Monoclonal Antibody [clone: E77]

Lieferant: Abcam
ABCAAB32399-100EA 672 EUR
ABCAAB32399-100
Anti-CCR8 Rabbit Monoclonal Antibody [clone: E77]
Antikörper
Type: Primary
Antigen: CCR8 (chemokine (C-C motif) receptor 8)
Clonality: monoclonal
Clone: E77
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR