Anti-KMT2A Mouse Monoclonal Antibody [clone: mmN4.4]
Lieferant: Abcam
ABCAAB32400-100EA 907 EUR
ABCAAB32400-100
Anti-KMT2A Mouse Monoclonal Antibody [clone: mmN4.4]
Antikörper
Type: Primary
Antigen: KMT2A (lysine (K)-specific methyltransferase 2A)
Clonality: monoclonal
Clone: mmN4.4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human