Anti-PI 3 Kinase p110 delta Rabbit Monoclonal Antibody [clone: Y387]

Lieferant: Abcam
ABCAAB32401-100EA 688 EUR
ABCAAB32401-100
Anti-PI 3 Kinase p110 delta Rabbit Monoclonal Antibody [clone: Y387]
Antikörper
Type: Primary
Antigen: PI 3 Kinase p110 delta
Clonality: monoclonal
Clone: Y387
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR