Anti-C Reactive Protein Rabbit Monoclonal Antibody [clone: Y284]

Lieferant: Abcam
ABCAAB32412-100EA 703 EUR
ABCAAB32412-100
Anti-C Reactive Protein Rabbit Monoclonal Antibody [clone: Y284]
Antikörper
Type: Primary
Antigen: C Reactive Protein
Clonality: monoclonal
Clone: Y284
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR