Anti-RPS6KA1 Rabbit Monoclonal Antibody [clone: E238]

Lieferant: Abcam
ABCAAB32413-100EA 703 EUR
ABCAAB32413-100
Anti-RPS6KA1 Rabbit Monoclonal Antibody [clone: E238]
Antikörper
Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: monoclonal
Clone: E238
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR