Anti-GTPase HRAS Rabbit Monoclonal Antibody [clone: Y132]

Lieferant: Abcam
ABCAAB32417-100EA 697 EUR
ABCAAB32417-100
Anti-GTPase HRAS Rabbit Monoclonal Antibody [clone: Y132]
Antikörper
Type: Primary
Antigen: GTPase HRAS
Clonality: monoclonal
Clone: Y132
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR