Anti-ATM Rabbit monoclonal antibody unconjugated [clone: Y170]

Lieferant: Abcam
ABCAAB32420-100EA 997 EUR
ABCAAB32420-100
Anti-ATM Rabbit monoclonal antibody unconjugated [clone: Y170]
Antikörper
Type: Primary
Antigen: ATM (ataxia telangiectasia mutated)
Clonality: monoclonal
Clone: Y170
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR