Anti-FABP7 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32423-100EA 935 EUR
ABCAAB32423-100
Anti-FABP7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP7 (fatty acid binding protein 7, brain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR