Anti-CBL Rabbit Monoclonal Antibody [clone: YE315]

Lieferant: Abcam
ABCAAB32425-100EA 688 EUR
ABCAAB32425-100
Anti-CBL Rabbit Monoclonal Antibody [clone: YE315]
Antikörper
Type: Primary
Antigen: CBL (Cbl proto-oncogene, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: YE315
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR