Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit monoclonal antibody unconjugated [clone: E308]

Lieferant: Abcam
ABCAAB32436-100EA 688 EUR
ABCAAB32436-100
Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit monoclonal antibody unconjugated [clone: E308]
Antikörper
Type: Primary
Antigen: Glutamate Receptor 1 (AMPA subtype)
Clonality: monoclonal
Clone: E308
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR