Anti-SIRT1 Rabbit Monoclonal Antibody [clone: E104]

Lieferant: Abcam
ABCAAB32441-100EA 763 EUR
ABCAAB32441-100
Anti-SIRT1 Rabbit Monoclonal Antibody [clone: E104]
Antikörper
Type: Primary
Antigen: SIRT1 (sirtuin 1)
Clonality: monoclonal
Clone: E104
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR