Anti-CDC25C Rabbit Monoclonal Antibody [clone: E302]

Lieferant: Abcam
ABCAAB32444-100EA 779 EUR
ABCAAB32444-100
Anti-CDC25C Rabbit Monoclonal Antibody [clone: E302]
Antikörper
Type: Primary
Antigen: CDC25C (cell division cycle 25C)
Clonality: monoclonal
Clone: E302
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR