Anti-MAP2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32454-100EA 980 EUR
ABCAAB32454-100
Anti-MAP2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP2 (microtubule-associated protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR