Anti-EIF4A3 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB32485-100EA 951 EUR
ABCAAB32485-100
Anti-EIF4A3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EIF4A3 (eukaryotic translation initiation factor 4A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat