Anti-CCNA2 Rabbit monoclonal antibody unconjugated [clone: E399]

Lieferant: Abcam
ABCAAB32498-100EA 688 EUR
ABCAAB32498-100
Anti-CCNA2 Rabbit monoclonal antibody unconjugated [clone: E399]
Antikörper
Type: Primary
Antigen: CCNA2 (cyclin A2)
Clonality: monoclonal
Clone: E399
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR