Anti-PRKCA Rabbit Monoclonal Antibody [clone: E195]
Lieferant: Abcam
ABCAAB32502-100EA 913 EUR
ABCAAB32502-100
Anti-PRKCA Rabbit Monoclonal Antibody [clone: E195]
Antikörper
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: monoclonal
Clone: E195
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen