Anti-BAX Rabbit monoclonal antibody unconjugated [clone: E63]
Lieferant: Abcam
ABCAAB32503-100EA 1020 EUR
ABCAAB32503-100
Anti-BAX Rabbit monoclonal antibody unconjugated [clone: E63]
Antikörper
Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: monoclonal
Clone: E63
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen