Anti-EIF2AK2 Rabbit monoclonal antibody unconjugated [clone: Y117]

Lieferant: Abcam
ABCAAB32506-100EA 779 EUR
ABCAAB32506-100
Anti-EIF2AK2 Rabbit monoclonal antibody unconjugated [clone: Y117]
Antikörper
Type: Primary
Antigen: EIF2AK2 (eukaryotic translation initiation factor 2-alpha kinase 2)
Clonality: monoclonal
Clone: Y117
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR