Anti-IRAK4 Rabbit Monoclonal Antibody [clone: Y279]

Lieferant: Abcam
ABCAAB32511-100EA 688 EUR
ABCAAB32511-100
Anti-IRAK4 Rabbit Monoclonal Antibody [clone: Y279]
Antikörper
Type: Primary
Antigen: IRAK4 (interleukin-1 receptor-associated kinase 4)
Clonality: monoclonal
Clone: Y279
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR