Anti-RB1 Rabbit Monoclonal Antibody [clone: E182]
Lieferant: Abcam
ABCAAB32513-100EA 990 EUR
ABCAAB32513-100
Anti-RB1 Rabbit Monoclonal Antibody [clone: E182]
Antikörper
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: monoclonal
Clone: E182
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen