Anti-PKA R2 Rabbit Monoclonal Antibody [clone: Y116]

Lieferant: Abcam
ABCAAB32514-100EA 688 EUR
ABCAAB32514-100
Anti-PKA R2 Rabbit Monoclonal Antibody [clone: Y116]
Antikörper
Type: Primary
Antigen: PKA R2
Clonality: monoclonal
Clone: Y116
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR