Anti-CREB Rabbit monoclonal antibody unconjugated [clone: E306]

Lieferant: Abcam
ABCAAB32515-100EA 779 EUR
ABCAAB32515-100
Anti-CREB Rabbit monoclonal antibody unconjugated [clone: E306]
Antikörper
Type: Primary
Antigen: CREB
Clonality: monoclonal
Clone: E306
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR