Anti-AIFM1 Rabbit Monoclonal Antibody [clone: E20]

Lieferant: Abcam
ABCAAB32516-100EA 763 EUR
ABCAAB32516-100
Anti-AIFM1 Rabbit Monoclonal Antibody [clone: E20]
Antikörper
Type: Primary
Antigen: AIFM1 (apoptosis-inducing factor, mitochondrion-associated, 1)
Clonality: monoclonal
Clone: E20
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR