Anti-MAP2K2 Rabbit Monoclonal Antibody [clone: Y78]

Lieferant: Abcam
ABCAAB32517-100EA 779 EUR
ABCAAB32517-100
Anti-MAP2K2 Rabbit Monoclonal Antibody [clone: Y78]
Antikörper
Type: Primary
Antigen: MAP2K2 (mitogen-activated protein kinase kinase 2)
Clonality: monoclonal
Clone: Y78
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR