Anti-Myosin Phosphatase 1+Myosin Phosphatase 2 Rabbit Monoclonal Antibody [clone: YE336]

Lieferant: Abcam
ABCAAB32519-100EA 703 EUR
ABCAAB32519-100
Anti-Myosin Phosphatase 1+Myosin Phosphatase 2 Rabbit Monoclonal Antibody [clone: YE336]
Antikörper
Type: Primary
Antigen: Myosin Phosphatase 1+Myosin Phosphatase 2
Clonality: monoclonal
Clone: YE336
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR