Anti-STAT6 Rabbit Monoclonal Antibody [clone: YE361]

Lieferant: Abcam
ABCAAB32520-100EA 749 EUR
ABCAAB32520-100
Anti-STAT6 Rabbit Monoclonal Antibody [clone: YE361]
Antikörper
Type: Primary
Antigen: STAT6 (signal transducer and activator of transcription 6, interleukin-4 induced)
Clonality: monoclonal
Clone: YE361
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR