Anti-PLEC Rabbit Monoclonal Antibody [clone: E398P]

Lieferant: Abcam
ABCAAB32528-100EA 688 EUR
ABCAAB32528-100
Anti-PLEC Rabbit Monoclonal Antibody [clone: E398P]
Antikörper
Type: Primary
Antigen: PLEC (plectin)
Clonality: monoclonal
Clone: E398P
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR