Anti-S6K Rabbit Monoclonal Antibody [clone: E343]

Lieferant: Abcam
ABCAAB32529-100EA 763 EUR
ABCAAB32529-100
Anti-S6K Rabbit Monoclonal Antibody [clone: E343]
Antikörper
Type: Primary
Antigen: S6K
Clonality: monoclonal
Clone: E343
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR