Anti-CHEK1 Rabbit Monoclonal Antibody [clone: E250]

Lieferant: Abcam
ABCAAB32531-100EA 688 EUR
ABCAAB32531-100
Anti-CHEK1 Rabbit Monoclonal Antibody [clone: E250]
Antikörper
Type: Primary
Antigen: CHEK1 (checkpoint kinase 1)
Clonality: monoclonal
Clone: E250
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR