Anti-LBR Rabbit Monoclonal Antibody [clone: E398L]

Lieferant: Abcam
ABCAAB32535-100EA 703 EUR
ABCAAB32535-100
Anti-LBR Rabbit Monoclonal Antibody [clone: E398L]
Antikörper
Type: Primary
Antigen: LBR (lamin B receptor)
Clonality: monoclonal
Clone: E398L
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR