Anti-MAPK3 Rabbit Monoclonal Antibody [clone: Y72]

Lieferant: Abcam
ABCAAB32537-100EA 779 EUR
ABCAAB32537-100
Anti-MAPK3 Rabbit Monoclonal Antibody [clone: Y72]
Antikörper
Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: monoclonal
Clone: Y72
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR