Anti-CASP9 Rabbit monoclonal antibody unconjugated [clone: E23]
Lieferant: Abcam
ABCAAB32539-100EA 1020 EUR
ABCAAB32539-100
Anti-CASP9 Rabbit monoclonal antibody unconjugated [clone: E23]
Antikörper
Type: Primary
Antigen: CASP9 (caspase 9, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: E23
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen