Anti-BTK Rabbit Monoclonal Antibody [clone: Y440]

Lieferant: Abcam
ABCAAB32555-100EA 672 EUR
ABCAAB32555-100
Anti-BTK Rabbit Monoclonal Antibody [clone: Y440]
Antikörper
Type: Primary
Antigen: BTK (Bruton agammaglobulinemia tyrosine kinase)
Clonality: monoclonal
Clone: Y440
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR