Anti-CCR9 Rabbit Monoclonal Antibody [clone: E99]

Lieferant: Abcam
ABCAAB32556-100EA 688 EUR
ABCAAB32556-100
Anti-CCR9 Rabbit Monoclonal Antibody [clone: E99]
Antikörper
Type: Primary
Antigen: CCR9 (chemokine (C-C motif) receptor 9)
Clonality: monoclonal
Clone: E99
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR