Anti-PTPN6 Rabbit Monoclonal Antibody [clone: Y476]

Lieferant: Abcam
ABCAAB32559-100EA 749 EUR
ABCAAB32559-100
Anti-PTPN6 Rabbit Monoclonal Antibody [clone: Y476]
Antikörper
Type: Primary
Antigen: PTPN6 (protein tyrosine phosphatase, non-receptor type 6)
Clonality: monoclonal
Clone: Y476
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR