Anti-EGFR Rabbit monoclonal antibody unconjugated [clone: E114]

Lieferant: Abcam
ABCAAB32562-100EA 749 EUR
ABCAAB32562-100
Anti-EGFR Rabbit monoclonal antibody unconjugated [clone: E114]
Antikörper
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: monoclonal
Clone: E114
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR