Anti-ACKR4 Rabbit Monoclonal Antibody [clone: E100]

Lieferant: Abcam
ABCAAB32564-100EA 626 EUR
ABCAAB32564-100
Anti-ACKR4 Rabbit Monoclonal Antibody [clone: E100]
Antikörper
Type: Primary
Antigen: ACKR4 (atypical chemokine receptor 4)
Clonality: monoclonal
Clone: E100
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR