Anti-ADAMTS9 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32565-100EA 666 EUR
ABCAAB32565-100
Anti-ADAMTS9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAMTS9 (ADAM metallopeptidase with thrombospondin type 1 motif, 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR