Anti-PRKDC Rabbit Monoclonal Antibody [clone: Y393]

Lieferant: Abcam
ABCAAB32566-100EA 779 EUR
ABCAAB32566-100
Anti-PRKDC Rabbit Monoclonal Antibody [clone: Y393]
Antikörper
Type: Primary
Antigen: PRKDC (protein kinase, DNA-activated, catalytic polypeptide)
Clonality: monoclonal
Clone: Y393
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR