Anti-MAPKAPK2 Rabbit Monoclonal Antibody [clone: E341]

Lieferant: Abcam
ABCAAB32567-100EA 981 EUR
ABCAAB32567-100
Anti-MAPKAPK2 Rabbit Monoclonal Antibody [clone: E341]
Antikörper
Type: Primary
Antigen: MAPKAPK2 (mitogen-activated protein kinase-activated protein kinase 2)
Clonality: monoclonal
Clone: E341
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR