Anti-Hsp90 beta Rabbit Monoclonal Antibody [clone: E296]

Lieferant: Abcam
ABCAAB32568-100EA 910 EUR
ABCAAB32568-100
Anti-Hsp90 beta Rabbit Monoclonal Antibody [clone: E296]
Antikörper
Type: Primary
Antigen: Hsp90 beta
Clonality: monoclonal
Clone: E296
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR