Anti-PI3 Kinase p110 beta Rabbit Monoclonal Antibody [clone: Y384]

Lieferant: Abcam
ABCAAB32569-100EA 749 EUR
ABCAAB32569-100
Anti-PI3 Kinase p110 beta Rabbit Monoclonal Antibody [clone: Y384]
Antikörper
Type: Primary
Antigen: PI3 Kinase p110 beta
Clonality: monoclonal
Clone: Y384
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR