Anti-PDGFRB Rabbit Monoclonal Antibody [clone: Y92]

Lieferant: Abcam
ABCAAB32570-100EA 792 EUR
ABCAAB32570-100
Anti-PDGFRB Rabbit Monoclonal Antibody [clone: Y92]
Antikörper
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: monoclonal
Clone: Y92
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR