Anti-PTK2B Rabbit Monoclonal Antibody [clone: YE353]

Lieferant: Abcam
ABCAAB32571-100EA 757 EUR
ABCAAB32571-100
Anti-PTK2B Rabbit Monoclonal Antibody [clone: YE353]
Antikörper
Type: Primary
Antigen: PTK2B (protein tyrosine kinase 2 beta)
Clonality: monoclonal
Clone: YE353
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR