Anti-CTNNB1 Rabbit monoclonal antibody unconjugated [clone: E247]

Lieferant: Abcam
ABCAAB32572-100EA 1020 EUR
ABCAAB32572-100
Anti-CTNNB1 Rabbit monoclonal antibody unconjugated [clone: E247]
Antikörper
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: monoclonal
Clone: E247
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR