Anti-MRP1 Mouse Monoclonal Antibody [clone: IU2H10]

Lieferant: Abcam
ABCAAB32574-100EA 688 EUR
ABCAAB32574-100
Anti-MRP1 Mouse Monoclonal Antibody [clone: IU2H10]
Antikörper
Type: Primary
Antigen: MRP1
Clonality: monoclonal
Clone: IU2H10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR