Anti-CAV1 Rabbit Monoclonal Antibody [clone: E249]

Lieferant: Abcam
ABCAAB32577-100EA 703 EUR
ABCAAB32577-100
Anti-CAV1 Rabbit Monoclonal Antibody [clone: E249]
Antikörper
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: monoclonal
Clone: E249
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR