Anti-KRT8 Rabbit Monoclonal Antibody [clone: E431-2]

Lieferant: Abcam
ABCAAB32579-100EA 713 EUR
ABCAAB32579-100
Anti-KRT8 Rabbit Monoclonal Antibody [clone: E431-2]
Antikörper
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: E431-2
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR