Anti-LMAN1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32583-100EA 682 EUR
ABCAAB32583-100
Anti-LMAN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LMAN1 (lectin, mannose-binding, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR