Anti-GABRA1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB32589-50EA 672 EUR
ABCAAB32589-50
Anti-GABRA1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GABRA1 (gamma-aminobutyric acid (GABA) A receptor, alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR